Skip to Content
Merck
All Photos(1)

Key Documents

804371

Sigma-Aldrich

Fro-DO, mixture of diastereomers

97%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$434.00

$434.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$434.00

About This Item

Empirical Formula (Hill Notation):
C24H22N2O6S2
Molecular Weight:
498.57
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$434.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

reaction suitability

core: nickel
reagent type: catalyst

SMILES string

O=C(N([C@H](C(C=CC=C1)=C1C2)[C@H]2C3)S3(=O)=O)/C=C/C(N4[C@H](C(C=CC=C5)=C5C6)[C@H]6CS4(=O)=O)=O

InChI

1S/C24H22N2O6S2/c27-21(25-23-17(13-33(25,29)30)11-15-5-1-3-7-19(15)23)9-10-22(28)26-24-18(14-34(26,31)32)12-16-6-2-4-8-20(16)24/h1-10,17-18,23-24H,11-14H2/b10-9+/t17-,18-,23+,24+/m1/s1

InChI key

CZVFUQRTDAQUQL-SKLQJEMHSA-N

Application

Enables the generation of quaternary centers by Ni-catalyzed cross coupling with aziridines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chung-Yang Dennis Huang et al.
Journal of the American Chemical Society, 137(17), 5638-5641 (2015-04-17)
A Ni-catalyzed Negishi cross-coupling with 1,1-disubstituted styrenyl aziridines has been developed. This method delivers valuable β-substituted phenethylamines via a challenging reductive elimination that affords a quaternary carbon. A novel electron-deficient olefin ligand, Fro-DO, proved crucial for achieving high rates and

Related Content

Doyle group focuses on nucleophilic fluorination using PyFluor and nickel catalysis with modular, air-stable nickel precatalyst.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service