Skip to Content
Merck
All Photos(1)

Key Documents

799629

Sigma-Aldrich

EncapGel-Cx Kit, chemically crosslinkable sIPN hydrogel kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$2,560.00

$2,560.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$2,560.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.23

$2,560.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Application

EncapGel-Cx Kit is a ready-to-use, chemically crosslinked hydrogel kit for the encapsulation and delivery of drugs, biomolecules, and cells. This hydrogel fuses the durability of synthetic polymers and the biorecognition of natural proteins to create a biocompatible delivery platform. The biodegradable matrix allows for temporally and spacially controlled delivery. Applications include:
  • Localized delivery of small molecules, proteins, nucleic acids, and cellular-based therapeutics
  • Tissue engineering
  • Regenerative medicine

Legal Information

ENCAPGEL is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service