Skip to Content
Merck
All Photos(1)

Documents

799610

Sigma-Aldrich

EncapGel-UV Kit, photo-crosslinkable sIPN hydrogel kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.23

Related Categories

Application

EncapGel-UV Kit is a ready-to-use, UV- crosslinked hydrogel kit for the encapsulation and delivery of drugs, biomolecules, and cells. This hydrogel fuses the durability of synthetic polymers and the biorecognition of natural proteins to create a biocompatible delivery platform. The biodegradable matrix allows for temporally and spacially controlled delivery. Applications include:
  • Localized delivery of small molecules, proteins, nucleic acids, and cellular-based therapeutics
  • Tissue engineering
  • Regenerative medicine

Legal Information

ENCAPGEL is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service