Skip to Content
Merck
All Photos(2)

Key Documents

78166

Sigma-Aldrich

N-(4-Phenylbenzylidene)benzylamine

≥98.0% (GC)

Synonym(s):

N-(4-Biphenylylmethylene)benzylamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$131.00

$131.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$131.00

About This Item

Empirical Formula (Hill Notation):
C20H17N
CAS Number:
Molecular Weight:
271.36
Beilstein:
3303002
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$131.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98.0% (GC)

form

powder

mp

58-62 °C (lit.)

functional group

amine
imine
phenyl

SMILES string

C(\N=C\c1ccc(cc1)-c2ccccc2)c3ccccc3

InChI

1S/C20H17N/c1-3-7-17(8-4-1)15-21-16-18-11-13-20(14-12-18)19-9-5-2-6-10-19/h1-14,16H,15H2/b21-16+

InChI key

MBJGFVKTDXQEPJ-LTGZKZEYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Indicator employed for the determination of the concentration of alkyllithium solutions[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L. Duhamel et al.
Journal of Organometallic Chemistry, 448, 1-1 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service