Skip to Content
Merck
All Photos(1)

Documents

231304

Sigma-Aldrich

N-Benzylidenebenzylamine

contains 100 ppm MEHQ as stabilizer, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N=CHC6H5
CAS Number:
Molecular Weight:
195.26
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

99%

form

liquid

contains

100 ppm MEHQ as stabilizer

refractive index

n20/D 1.6004 (lit.)

bp

143-144 °C/5 mmHg (lit.)

density

1.038 g/mL at 25 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

C(\N=C\c1ccccc1)c2ccccc2

InChI

1S/C14H13N/c1-3-7-13(8-4-1)11-15-12-14-9-5-2-6-10-14/h1-11H,12H2/b15-11+

InChI key

MIYKHJXFICMPOJ-RVDMUPIBSA-N

Related Categories

Application

Indicator for organolithium assay.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service