Skip to Content
Merck
All Photos(1)

Documents

B34680

Sigma-Aldrich

Biphenyl-4-carboxaldehyde

99%

Synonym(s):

4-Phenylbenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4CHO
CAS Number:
Molecular Weight:
182.22
Beilstein:
606693
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

crystals

bp

184 °C/11 mmHg (lit.)

mp

57-59 °C (lit.)

SMILES string

O=Cc1ccc(cc1)-c2ccccc2

InChI

1S/C13H10O/c14-10-11-6-8-13(9-7-11)12-4-2-1-3-5-12/h1-10H

InChI key

ISDBWOPVZKNQDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

352.0 - 381.9 °F

Flash Point(C)

177.8 - 194.4 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Mariela M Nolasco et al.
Molecules (Basel, Switzerland), 25(6) (2020-03-22)
The present work emphasizes the value of periodic density functional theory (DFT) calculations in the assessment of the vibrational spectra of molecular crystals. Periodic calculations provide a nearly one-to-one match between the calculated and observed bands in the inelastic neutron
Yuki Okabe et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 22(5), 713-725 (2017-01-14)
The construction of molecular catalysts that are active toward CO
Danuta Sek et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 175, 24-35 (2016-12-25)
The new Schiff bases bearing anthracene unit were synthesized from 2-aminoanthracene and various aldehydes such as: benzaldehyde, 4-(diphenylamino)benzaldehyde, 9-phenanthrenecarboxaldehyde, 9-anthracenecarboxaldehyde, and biphenyl-4-carboxaldehyde, 2-naphthaldehyde. Resulted azomethines were characterized by IR, NMR (
Sorel Jatunov et al.
Carbohydrate polymers, 123, 288-296 (2015-04-07)
A variety of fluorescent imino and secondary amino chitosans were synthesized under very mild conditions by reaction of the biopolymer amino functions with aromatic aldehydes in an acidified methanolic suspension. Simultaneous reactions of several aldehydes with chitosan were successfully carried

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service