Skip to Content
Merck
All Photos(1)

Key Documents

776998

Sigma-Aldrich

(R)-1,1,1-Trifluoro-2-propanol

≥95%

Synonym(s):

(R)-1,1,1-Trifluoropropan-2-ol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$173.00

$173.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$173.00

About This Item

Empirical Formula (Hill Notation):
C3H5F3O
CAS Number:
Molecular Weight:
114.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$173.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

liquid

optical activity

[α]22/D +8.3°, c = 1% in chloroform

functional group

fluoro
hydroxyl

storage temp.

2-8°C

SMILES string

C[C@@H](O)C(F)(F)F

InChI

1S/C3H5F3O/c1-2(7)3(4,5)6/h2,7H,1H3/t2-/m1/s1

InChI key

GILIYJDBJZWGBG-UWTATZPHSA-N

Related Categories

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F

Flash Point(C)

18 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service