Skip to Content
Merck
All Photos(1)

Key Documents

776165

Sigma-Aldrich

1-Bromo-2-(trifluoromethoxy)ethane

Synonym(s):

2-Bromoethyl trifluoromethyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$251.00

$251.00


Estimated to ship onApril 14, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$251.00

About This Item

Empirical Formula (Hill Notation):
C3H4BrF3O
CAS Number:
Molecular Weight:
192.96
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$251.00


Estimated to ship onApril 14, 2025


Request a Bulk Order

form

liquid

Quality Level

refractive index

n20/D 1.367

density

1.674 g/mL at 25 °C

functional group

bromo
fluoro

storage temp.

2-8°C

SMILES string

BrCCOC(F)(F)F

InChI

1S/C3H4BrF3O/c4-1-2-8-3(5,6)7/h1-2H2

InChI key

YLLDCQMJIXHFQZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service