Skip to Content
Merck
All Photos(1)

Key Documents

540323

Sigma-Aldrich

1,1,1-Trifluoro-2-propanol

97%

Synonym(s):

1,1,1-Trifluoroisopropanol, 1,1,1-Trifluoropropane-2-ol, 1-(Trifluoromethyl)ethanol, 1-Methyl-2,2,2-trifluoroethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CH(OH)CH3
CAS Number:
Molecular Weight:
114.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.316 (lit.)

bp

81-82 °C (lit.)

density

1.259 g/mL at 25 °C (lit.)

functional group

fluoro
hydroxyl

SMILES string

CC(O)C(F)(F)F

InChI

1S/C3H5F3O/c1-2(7)3(4,5)6/h2,7H,1H3

InChI key

GILIYJDBJZWGBG-UHFFFAOYSA-N

General description

1,1,1-Trifluoro-2-propanol, a fluoroalcohol, is an organic solvent. Vibrational spectral analysis of OH and OD groups of 1,1,1-trifluoro-2-propanol and its deuterated derivatives has been conducted.

Application

1,1,1-Trifluoro-2-propanol may be used in the synthesis of 1,1,1-trifluoropropan-2-yl 2-(4-hydroxy-3-methoxyphenyl)acetate and 1,1,1-trifluoropropan-2-yl 2-(3-ethoxy-4-hydroxyphenyl)acetate.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.9 °F - closed cup

Flash Point(C)

18.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthetic mammalian prions.
Legname G, et al.
Science, 305, 673-676 (2004)
Fluoroalcohols-XXI. Infrared, matrix infrared and Raman spectra of 1, 1, 1-trifluoro-2-propanol and its three deuterated analogues.
Murto J, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 31(4), 479-493 (1975)
Heng Zhang et al.
PloS one, 9(5), e96518-e96518 (2014-05-07)
Soft drugs are molecules that are purposefully designed to be rapidly metabolized (metabolically labile). In anesthesia, the soft drug is useful because it enables precise titration to effect and rapid recovery, which might allow swift and clear-headed recovery of consciousness

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service