Skip to Content
Merck
All Photos(1)

Key Documents

639842

Sigma-Aldrich

1-Bromo-2-(2-methoxyethoxy)ethane

95%

Synonym(s):

1-(2-Bromoethoxy)-2-methoxyethane, 1-Bromo-2-[1-[2-(methyloxy)ethyl]oxy]ethane, 1-Bromo-3,6-dioxaheptane, 2-(2-Methoxyethoxy)ethyl bromide, 2-(2′-Methoxyethoxy)ethyl bromide, 2-Bromoethyl 2-methoxyethyl ether, 7-Bromo-2,5-dioxaheptane

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$364.00
50 G
$1,810.00

$364.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$364.00
50 G
$1,810.00

About This Item

Linear Formula:
CH3(OCH2CH2)2Br
CAS Number:
Molecular Weight:
183.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$364.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

refractive index

n20/D 1.455 (lit.)

bp

156 °C (lit.)
37-40 °C/0.01 mmHg (lit.)

density

1.3600 g/mL at 25 °C (lit.)

functional group

bromo
ether

storage temp.

2-8°C

SMILES string

COCCOCCBr

InChI

1S/C5H11BrO2/c1-7-4-5-8-3-2-6/h2-5H2,1H3

InChI key

HUXJXNSHCKHFIL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1-Bromo-2-(2-methoxyethoxy)ethane can be used in the synthesis of 1,4-diketo-3,6-diphenylpyrrolo[3,4-c]pyrrole(DPP)derivatives, which are stable and efficient pigments for two-photon excited fluorescence microscopy.[1] It can also be used in the synthesis of a squarine dye, which exhibits a selective fluorogenic response towards zinc cation.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.9 °F - closed cup

Flash Point(C)

68.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Design and synthesis of a squaraine based near-infrared fluorescent probe for the ratiometric detection of Zn 2+ ions"
Huang Y, et al.
Dyes and Pigments, 99(03), 699- 704 (2013)
"Water-soluble diketopyrrolopyrrole derivatives for two-photon excited fluorescence microscopy"
Ftouni H, et al.
Dyes and Pigments, 97(01), 77-83 (2013)

Questions

  1. Ist dieses Produkt mit Na2CO3 stabilisiert?

    1 answer
    1. This product does not contain stabilizer.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service