Skip to Content
Merck
All Photos(1)

Key Documents

77004

Sigma-Aldrich

Tetrabutylammonium hydrogen difluoride solution

~50% in acetonitrile

Synonym(s):

TBABF, Tetrabutylammonium bifluoride, Tetrabutylammonium difluoride

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$860.00

$860.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$860.00

About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(HF2)
CAS Number:
Molecular Weight:
281.47
Beilstein:
4729304
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$860.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

form

solution

concentration

~50% in acetonitrile

impurities

≤1% water

functional group

amine

storage temp.

2-8°C

SMILES string

[F-].F[H].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.2FH/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;;/h5-16H2,1-4H3;2*1H/q+1;;/p-1

InChI key

ZHBDKVWQJKYIFF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Used as an etchant for silica films

Reactant for:
  • Nucleophilic fluorination
  • Ring-opening fluorination reaction
  • Substitution reactions

Other Notes

Efficient fluorinating agent[1][2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

<50.0 °F - closed cup

Flash Point(C)

< 10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Bosch et al.
Tetrahedron Letters, 28, 4733-4733 (1987)
F. Camps et al.
The Journal of Organic Chemistry, 54, 4294-4294 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service