Skip to Content
Merck
All Photos(3)

Documents

441457

Sigma-Aldrich

Tetrabutylammonium difluorotriphenylsilicate

97%

Synonym(s):

TBAT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N[(C6H5)3SiF2]
CAS Number:
Molecular Weight:
539.86
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

157-163 °C (lit.)

SMILES string

CCCC[N+](CCCC)(CCCC)CCCC.F[Si-](F)(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C18H15F2Si.C16H36N/c19-21(20,16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4/h1-15H;5-16H2,1-4H3/q-1;+1

InChI key

RQBKGJOQACIQDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrabutylammonium difluorotriphenylsilicate (TBAT) is a non-hygroscopic, organic soluble fluoride source and it is very effective for nucleophilic displacement reactions. Furthermore, TBAT is used as a catalytic promoter.

Application

A fluoride source for nucleophilic fluorination.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kohei Fuchibe et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(12), 2831-2838 (2016-12-03)
Silyl dienol ethers, prepared from α,β-unsaturated ketones, underwent proton sponge-catalyzed difluorocyclopropanation with trimethylsilyl 2,2-difluoro-2-fluorosulfonylacetate in a regioselective manner, leading to 1,1-difluoro-2-siloxy-2-vinylcyclopropanes in good yields. The cyclopropanes thus obtained were in turn subjected to fluoride-ion-catalyzed ring opening to afford 1-fluorovinyl vinyl
Yansheng Wang et al.
Oncology reports, 38(3), 1579-1586 (2017-07-18)
The aim of this study was to elucidate the effects of iodine-131 on the induction of apoptosis in human cardiac muscle cells and the underlying molecular mechanisms. We found that iodine-131 reduced cell proliferation, induced apoptosis, induced p53, PIDD, t-BID
Tetrabutylammonium Difluorotriphenylsilicate (TBAT )
Simpkins NS, et al.
Encyclopedia of Reagents for Organic Synthesis, Second Edition, 1-14 (2001)
Nucleophilic addition of TMSCCl3 to N-phosphinoyl benzaldimines: a route to N-phosphinoyl-?-(trichloromethyl)benzylamines
Wahl B, et al.
Tetrahedron Letters, 55(42), 5829-5831 (2014)
Journal of the American Chemical Society, 117, 5166-5166 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service