Skip to Content
Merck
All Photos(1)

Key Documents

763012

Sigma-Aldrich

CPhos Pd G2

95%

Synonym(s):

CPhos-G2-Palladacycle, CPhos-Pd-G2, Chloro[(2-dicyclohexylphosphino-2′,6′-bis(N,N-dimethylamino)-1,1′-biphenyl)-2-(2′-amino-1,1′-biphenyl)]palladium(II)

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$355.00

$355.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$355.00

About This Item

Empirical Formula (Hill Notation):
C40H51ClN3PPd
CAS Number:
Molecular Weight:
746.70
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$355.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

95%

form

solid

feature

generation 2

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

152-163 °C (decomposition)

functional group

phosphine

SMILES string

Nc1ccccc1-c2ccccc2[Pd]Cl.CN(C)c3cccc(N(C)C)c3-c4ccccc4P(C5CCCCC5)C6CCCCC6

InChI

1S/C28H41N2P.C12H10N.ClH.Pd/c1-29(2)25-19-13-20-26(30(3)4)28(25)24-18-11-12-21-27(24)31(22-14-7-5-8-15-22)23-16-9-6-10-17-23;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;;/h11-13,18-23H,5-10,14-17H2,1-4H3;1-6,8-9H,13H2;1H;/q;;;+1/p-1

InChI key

DHXLRDVUKOAYPC-UHFFFAOYSA-M

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Buchwald precatalysts and ligands are bulky electron-rich dialkylbiaryl phospine-based catalysts and structurally related ligands that improve reactivity in Pd-catalyzed cross - coupling reactions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service