Skip to Content
Merck
All Photos(2)

Key Documents

756504

Sigma-Aldrich

Cerium magnesium aluminate, terbium doped

greener alternative

99% trace metals basis

Synonym(s):

Aluminum cerium magnesium terbium oxide, Green phosphor, Terbium doped aluminum cerium magnesium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ce0.63Tb0.37MgAl11O19
CAS Number:
Molecular Weight:
772.17
UNSPSC Code:
12352300
NACRES:
NA.23

Assay

99% trace metals basis

form

powder

composition

Ce0.63Tb0.37MgAl11O19

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

4-8 μm (d50)

mp

>1000 °C (lit.)

fluorescence

λex 544 nm±2 nm (UV excitation source)

greener alternative category

General description

Cerium magnesium aluminate, terbium doped is a phosphor material, which has a magnetoplumbite structure and luminesces effectively in the UV region. The addition of terbium allows the transfer of excitation energy from Ce3+ to Tb3+, resulting in the formation of green luminescence. It can be prepared by methods such as spray pyrolysis and combustion method.[1]
Color coordinate (x): 0.335 +/- 0.015

Color coordinate (y): 0.590 +/- 0.015
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Cerium magnesium aluminate, terbium doped can be used in the development of display devices such as plasma display panels (PDPs), field emission displays (FEDs), and electroluminescence panels.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li, Y.J.; Kwon, Y.W.; Jones, M.; et al.
Semiconductor Science and Technology, 20, 720-720 (2005)
Photoluminescence properties of Ce1-xTbxMgAl11O19 phosphor particles prepared by spray pyrolysis
Kang YC, et al.
Japanese Journal of Applied Physics, 38(4R), 2013-2013 (1999)
Hong, S.K.; Jung, D.S.; Ju, S.H.; Koo, H.Y.; Kang, Y.C.
Journal of Materials Science: Materials in Electronics, 17, 341-341 (2006)

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service