Skip to Content
Merck
All Photos(1)

Key Documents

752134

Sigma-Aldrich

Cyclohexylmagnesium chloride solution

1.3 M in THF/toluene (1:1)

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$98.40

$98.40


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$98.40

About This Item

Empirical Formula (Hill Notation):
C6H11ClMg
CAS Number:
Molecular Weight:
142.91
Beilstein:
3535440
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$98.40


Estimated to ship onMay 30, 2025


Request a Bulk Order

form

solution

reaction suitability

reaction type: Grignard Reaction

concentration

1.3 M in THF/toluene (1:1)

density

0.938 g/mL at 25 °C

SMILES string

Cl[Mg]C1CCCCC1

InChI

1S/C6H11.ClH.Mg/c1-2-4-6-5-3-1;;/h1H,2-6H2;1H;/q;;+1/p-1

InChI key

WMJMABVHDMRMJA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F

Flash Point(C)

-11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service