Skip to Content
Merck
All Photos(1)

Key Documents

740861

Sigma-Aldrich

1,3,5-Triaza-7-phosphaadamantane-7-oxide

97%

Synonym(s):

1,3,5-Triaza-7-phosphatricyclo[3.3.1.13,7]decane, 7-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N3OP
CAS Number:
Molecular Weight:
173.15
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

280-290 °C

storage temp.

2-8°C

SMILES string

O=P12CN3CN(CN(C3)C1)C2

InChI

1S/C6H12N3OP/c10-11-4-7-1-8(5-11)3-9(2-7)6-11/h1-6H2

InChI key

BPHLLGJNQPYNDP-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service