Skip to Content
Merck
All Photos(1)

Key Documents

326666

Sigma-Aldrich

Palladium

Palladium

powder, <1 μm, ≥99.9% trace metals basis

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$485.00
5 G
$2,070.00

$485.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$485.00
5 G
$2,070.00

About This Item

Linear Formula:
Pd
CAS Number:
Molecular Weight:
106.42
EC Number:
MDL number:
UNSPSC Code:
12141733
PubChem Substance ID:
NACRES:
NA.23

$485.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99.9% trace metals basis

form

powder

resistivity

9.96 μΩ-cm, 20°C

particle size

<1 μm

bp

2970 °C (lit.)

mp

1554 °C (lit.)

density

12.02 g/cm3 (lit.)

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Average particle size determined by Fisher sub-sieve sizer

Application


  • A Novel Photosensitizer Based 450-nm Blue Laser-Mediated Photodynamic Therapy Induces Apoptosis in Colorectal Cancer - in Vitro and in Vivo Study.: This study demonstrates the effectiveness of a 450-nm light source in photodynamic therapy, showing significant apoptosis in colorectal cancer cells both in vitro and in vivo (Zeng J et al., 2024).



Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefano Nicolai et al.
The Journal of organic chemistry, 78(8), 3783-3801 (2013-03-21)
Tetrahydrofurans and pyrrolidines are among the most important heterocycles found in bioactive compounds. Cyclization-functionalization domino reactions of alcohols or amines onto olefins constitute one of the most efficient methods to access them. In this context, oxy- and aminoalkynylation are especially
Xuxing Chen et al.
Organic & biomolecular chemistry, 11(16), 2582-2585 (2013-03-19)
A one-pot two-step synthesis of versatile indenones has been developed. This palladium(II)-catalyzed transformation involves generation and condensation of ortho-functionalized 1,2-benzils from 2-(2-arylethynylphenyl)acetonitriles using Ph2SO as the oxidant. The resulting 3-cyanoindenones can be converted to various valuable molecules.
Sandeep N Raikar et al.
The Journal of organic chemistry, 78(8), 3832-3846 (2013-03-26)
Cu-catalyzed three-component coupling of imines with benzoyl chloride and terminal arylalkynes followed by enyne ring-closing metathesis (RCM) and Heck cyclization afforded medicinally relevant benzoindolines, cyclopropane-fused indenopyridines, pyrroloquinolines, or 1,7-tetrahydrophenanthrolines via divergent cyclization pathways. Unexpectedly, the Pd-catalyzed cyclization of heterocyclic dienes
Meng Wang et al.
Organic & biomolecular chemistry, 11(16), 2574-2577 (2013-03-15)
A novel Pd/Cu catalyzed domino reaction for the synthesis of functionalized pyrrolo[1,2-b]pyridazines from readily accessible (hetero)aryl propargyl alcohols and 1-amino-2-bromopyrroles was developed. This cascade process involves a Sonogashira cross-coupling reaction, an isomerization and an intramolecular condensation.
Ling Li et al.
Nature chemistry, 5(7), 607-612 (2013-06-22)
The development of transition metal-catalysed cross-coupling reactions has greatly influenced the manner in which the synthesis of complex organic molecules is approached. A wide variety of methods are now available for the formation of C(sp(2))-C(sp(2)) bonds, and more recent work

Articles

Permanent magnets are an essential technology for energy conversion. Motors and generators are used to convert energy between electrical and mechanical forms.

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service