Skip to Content
Merck
All Photos(1)

Key Documents

705810

Sigma-Aldrich

3,6-Dichloro-2-hydroxybenzoic acid-(phenyl-13C6)

99 atom % 13C, 98% (CP)

Synonym(s):

3,6-Dichlorosalicylic acid-(ring-13C6)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$3,080.00
10 MG
$5,550.00

$3,080.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request more information

Select a Size

Change View
5 MG
$3,080.00
10 MG
$5,550.00

About This Item

Empirical Formula (Hill Notation):
13C6CH4Cl2O3
CAS Number:
Molecular Weight:
212.97
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

$3,080.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request more information

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+6

storage temp.

−20°C

SMILES string

OC(=O)[13c]1[13c](Cl)[13cH][13cH][13c](Cl)[13c]1O

InChI

1S/C7H4Cl2O3/c8-3-1-2-4(9)6(10)5(3)7(11)12/h1-2,10H,(H,11,12)/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

FKIKPQHMWFZFEB-IDEBNGHGSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service