Skip to Content
Merck
All Photos(1)

Key Documents

728187

Sigma-Aldrich

1-Phenyl-2-propyn-1-one

≥95.0% (HPLC)

Synonym(s):

1-Phenylpropynone, Ethynyl phenyl ketone

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$356.00

$356.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$356.00

About This Item

Empirical Formula (Hill Notation):
C9H6O
CAS Number:
Molecular Weight:
130.14
Beilstein:
606216
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

$356.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95.0% (HPLC)

form

lumps

storage temp.

2-8°C

SMILES string

O=C(C#C)c1ccccc1

InChI

1S/C9H6O/c1-2-9(10)8-6-4-3-5-7-8/h1,3-7H

InChI key

JITPLZPWKYUTDM-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Jaenicke et al.
European journal of biochemistry, 138(2), 319-325 (1984-01-16)
3-Oxo-3-phenylpropyne and 3-oxo-3-phenylpropene were synthesized as active-site-directed irreversible inhibitors of the bitter almond hydroxynitrile lyase (EC 4.1.2.10), an FAD-protein. The substrate and competitors (e.g. benzoate) decrease the rate of the inhibitor-mediated deactivation of the enzyme. By excess addition of either
Unsaturated lactones; researches on acetylenic compounds; a new route to growth-inhibitory alpha beta-ethylenic gamma- and delta-lactones.
L J HAYNES et al.
Journal of the Chemical Society, 954-957 (1946-10-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service