Skip to Content
Merck
All Photos(2)

Key Documents

78976

Sigma-Aldrich

(R)-1-Phenyl-2-propyn-1-ol

≥99.0% (sum of enantiomers, GC)

Synonym(s):

(R)-α-Ethynylbenzyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$358.00

$358.00


Estimated to ship onApril 22, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$358.00

About This Item

Empirical Formula (Hill Notation):
C9H8O
CAS Number:
Molecular Weight:
132.16
Beilstein:
3195857
MDL number:
UNSPSC Code:
12352003
PubChem Substance ID:
NACRES:
NA.22

$358.00


Estimated to ship onApril 22, 2025


Request a Bulk Order

Assay

≥99.0% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D −28±2°, c = 3.2% in chloroform

optical purity

enantiomeric ratio: ≥96:4 (HPLC)

refractive index

n20/D 1.551

density

1.067 g/mL at 20 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

O[C@H](C#C)c1ccccc1

InChI

1S/C9H8O/c1-2-9(10)8-6-4-3-5-7-8/h1,3-7,9-10H/t9-/m1/s1

InChI key

UIGLAZDLBZDVBL-SECBINFHSA-N

Caution

may discolor to yellow during storage

Other Notes

Chiral building block; synthesis of substituted allenes[1][2][3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C.J. Elsevier et al.
The Journal of Organic Chemistry, 47, 2194-2194 (1982)
C.J. Elsevier et al.
The Journal of Organic Chemistry, 54, 3726-3726 (1989)
The Journal of Organic Chemistry, 48, 1103-1103 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service