Skip to Content
Merck
All Photos(1)

Key Documents

78960

Sigma-Aldrich

Phenyl propargyl ether

technical, ≥90% (GC)

Synonym(s):

3-Phenoxy-1-propyne

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$175.00

$175.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
$175.00

About This Item

Linear Formula:
HC≡CCH2OC6H5
CAS Number:
Molecular Weight:
132.16
Beilstein:
2040910
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$175.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical

Assay

≥90% (GC)

form

liquid

refractive index

n20/D 1.535

density

1.030 g/mL at 20 °C (lit.)

functional group

phenoxy

storage temp.

2-8°C

SMILES string

C#CCOc1ccccc1

InChI

1S/C9H8O/c1-2-8-10-9-6-4-3-5-7-9/h1,3-7H,8H2

InChI key

AIQRJSXKXVZCJO-UHFFFAOYSA-N

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service