Skip to Content
Merck
All Photos(1)

Key Documents

727911

Sigma-Aldrich

1-Ethyl-3-methylimidazolium aminoacetate

≥96% (HPLC and enzymatic)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15N3O2
CAS Number:
Molecular Weight:
185.22
Beilstein:
9953832
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96% (HPLC and enzymatic)

form

liquid

impurities

≤0.5% halogenides /chloride, bromide, iodide)
≤10% water

SMILES string

NCC([O-])=O.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.C2H5NO2/c1-3-8-5-4-7(2)6-8;3-1-2(4)5/h4-6H,3H2,1-2H3;1,3H2,(H,4,5)/q+1;/p-1

InChI key

DVTRRYWFKKZCIE-UHFFFAOYSA-M

Application

1-Ethyl-3-methylimidazolium aminoacetate (1-ethyl-3-methylimidazolium glycinate) is an amino acid-based ionic liquid, which can be used:
  • To prepare amino acid immobilized porous supported materials as efficient adsorbents to capture CO2.
  • As an extractant for the separation of phenolic compounds.
  • As a catalyst in the aza-Michael reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The essential role of hydrogen-bonding interaction in the extractive separation of phenolic compounds by ionic liquid
Yang Q, et al.
AIChE Journal, 59(5), 1657-1667 (2013)
Ionic amino acids: Application as organocatalysts in the aza-Michael reaction
Morimoto N, et al.
J. Mol. Catal. A: Chem., 368(5), 31-37 (2013)
Immobilization of amino acid ionic liquids into nanoporous microspheres as robust sorbents for CO2 capture
Wang X, et al.
Journal of Material Chemistry A, 1(9), 2978-2982 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service