Skip to Content
Merck
All Photos(1)

Key Documents

272841

Sigma-Aldrich

1-Ethyl-3-methylimidazolium chloride

98%

Synonym(s):

EMIC

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$59.60
25 G
$194.00

$59.60

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 23, 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
$59.60
25 G
$194.00

About This Item

Empirical Formula (Hill Notation):
C6H11ClN2
CAS Number:
Molecular Weight:
146.62
Beilstein:
5163190
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$59.60

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 23, 2025Details


Request a Bulk Order

Quality Level

Assay

98%

form

solid

mp

77-79 °C (lit.)

SMILES string

[Cl-].CCn1cc[n+](C)c1

InChI

1S/C6H11N2.ClH/c1-3-8-5-4-7(2)6-8;/h4-6H,3H2,1-2H3;1H/q+1;/p-1

InChI key

BMQZYMYBQZGEEY-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

1-Ethyl-3-methylimidazolium chloride is an organic salt and a room-temperature ionic liquid (RTIL). It is used as a solvent in many organic reactions due to its unique properties like small vapor pressures, high thermal stabilities and ionic conductivities.[1]

1-Ethyl-3-methylimidazolium chloride is an imidazolium-based ionic liquid used as a solvent in the Diels−Alder Reaction.[2]

Application

EMIC may be used in the preparation of aluminum chloride-EMIC,[3] zinc chloride-EMIC[4] and EMIC /tetrafluoroborate molten salts, which are useful in electrodeposition studies.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

370.4 °F - closed cup

Flash Point(C)

188 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrodeposition of aluminum on magnesium alloy in aluminum chloride (AlCl 3)-1-ethyl-3-methylimidazolium chloride (EMIC) ionic liquid and its corrosion behavior.
Chang JK, et al.
Electrochemical Communications, 9(7), 1602-1606 (2007)
Carlos Rey-Castro et al.
The journal of physical chemistry. B, 110(29), 14426-14435 (2006-07-21)
We present here equilibrium molecular dynamics simulation results for self-diffusion coefficients, shear viscosity, and electrical conductivity in a model ionic liquid (1-ethyl-3-methylimidazolium chloride) at different temperatures. The Green-Kubo relations were employed to evaluate the transport coefficients. When compared with available
Lewis acidity dependency of the electrochemical window of zinc chloride-1-ethyl-3-methylimidazolium chloride ionic liquids.
Hsiu SI, et al.
Electrochimica Acta, 47(27), 4367-4372 (2002)
Elucidation of rate variations for a Diels- Alder reaction in ionic liquids from QM/MM simulations
Acevedo O, et al.
Journal of chemical theory and computation, 3(1), 132-138 (2007)
Anshu Sharma et al.
Carbohydrate polymers, 134, 617-626 (2015-10-03)
Ionic liquids (IL) can alter the physical properties of agar hydrogels. Rheology studies show that gels with wide range of storage moduli (gel strength) G0 values ranging from 1 to 20 KPa could be made in imidazolium based IL solutions

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service