Skip to Content
Merck
All Photos(1)

Documents

727709

Sigma-Aldrich

1-Allyl-3-methylimidazolium dicyanamide

≥98.5% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11N5
CAS Number:
Molecular Weight:
189.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.5% (HPLC)

form

liquid

impurities

≤1.0% water

SMILES string

N#C[N-]C#N.C[n+]1ccn(CC=C)c1

InChI

1S/C7H11N2.C2N3/c1-3-4-9-6-5-8(2)7-9;3-1-5-2-4/h3,5-7H,1,4H2,2H3;/q+1;-1

InChI key

LAYIDOPTTZCESY-UHFFFAOYSA-N

General description

1-Allyl-3-methylimidazolium dicyanamide is a 1-allyl-3-methylimidazolium-based ionic liquid1 that can be prepared by reacting 1-allyl-3-methylimidazoliumchloride with sodium dicyanamide.2

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Theoretical study on cation-anion interaction and vibrational spectra of 1-allyl-3-methylimidazolium-based ionic liquids.
Xuan X, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 78(5), 1492-1499 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service