Skip to Content
Merck
All Photos(1)

Key Documents

724424

Sigma-Aldrich

1-Ethyl-3-methylimidazolium tetrachloroaluminate

Synonym(s):

1-Ethyl-3-methyl-1H-imidazolium tetrachloroaluminate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11AlCl4N2
CAS Number:
Molecular Weight:
279.96
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

liquid

impurities

≤1% H2O

refractive index

n20/D 1.484

mp

9 °C

density

1.304 g/mL at 25 °C

application(s)

battery manufacturing

SMILES string

Cl[Al-](Cl)(Cl)Cl.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.Al.4ClH/c1-3-8-5-4-7(2)6-8;;;;;/h4-6H,3H2,1-2H3;;4*1H/q+1;+3;;;;/p-4

InChI key

UYYXEZMYUOVMPT-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Dye sensitized solar cell, capacitor technology, batteries[1][2][3]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

424.4 °F

Flash Point(C)

218 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Dai, Qing; MacFarlane, Douglas R.; Forsyth, Maria
Solid State Ionics, 177, 395-395 (2006)
Carlin, Richard T.; De Long, Hugh C.; Fuller, Joan; Trulove, Paul C.
Journal of the Electrochemical Society, 141, L73-L73 (1994)
Amir, N.; Vestfrid, Y.; Chusid, O.; Gofer, Y.; Aurbach, D.
Journal of Power Sources, 174, 1234-1234 (2007)

Articles

Ionic liquids, also called room temperature ionic liquids, are organic salts that are liquid at, or close to, room temperature.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

The critical technical challenges associated with the commercialization of electric vehicle batteries include cost, performance, abuse tolerance, and lifespan.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service