1-Ethyl-3-methylimidazolium tetrachloroaluminate
Synonym(s):
1-Ethyl-3-methyl-1H-imidazolium tetrachloroaluminate
Empirical Formula (Hill Notation):
C6H11AlCl4N2
CAS Number:
Molecular Weight:
279.96
Products
Brand
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing