Skip to Content
Merck
All Photos(5)

Key Documents

809942

Sigma-Aldrich

Dimethyl carbonate

greener alternative

battery grade, ≥99.9%, acid <10 ppm, H2O <10 ppm

Synonym(s):

DMC, dimethyl ester, Carbonic acid dimethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$353.00
500 G
$1,330.00

$353.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$353.00
500 G
$1,330.00

About This Item

Linear Formula:
(CH3O)2CO
CAS Number:
Molecular Weight:
90.08
Beilstein:
635821
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.23

$353.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

battery grade

Quality Level

vapor density

3.1 (vs air)

vapor pressure

18 mmHg ( 21.1 °C)

Assay

≥99.9%

form

liquid

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Safer Solvents and Auxiliaries
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤10 ppm H2O
≤10 ppm acid

refractive index

n20/D 1.368 (lit.)

bp

90 °C (lit.)

mp

2-4 °C (lit.)

density

1.069 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

O=C(OC)OC

InChI

1S/C3H6O3/c1-5-3(4)6-2/h1-2H3

InChI key

IEJIGPNLZYLLBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is used as a green chemical. Click here for more information.

Application

Alkyl carbonates find applications as solvents for lithium ion battery electrolytes and the use of high quality battery grade electrolytes having extremely low water (<10 ppm) and acid (<10 ppm) contents are critical for achieving high electrochemical performance.

Other Notes

  • These additives have low water content (less than 100 ppm).
  • Please handle under inert and moisture free environment (glove box).
  • Keep containers tightly closed.
  • Keep away from heat and ignition sources.
  • Store in a cool and dry place.
  • Avoid storing together with oxidizers.

Legal Information

Product of MU Ionic Solutions Corp

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Analysis of carbonate solvents in lithium-ion batteries like ethylene carbonate, dimethyl carbonate, diethyl carbonate, and more using SPB®-624 column.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Due to the adverse impact of the continued use of fossil fuels on the earth’s environment and climate, researchers have been asked to develop new approaches for producing power using renewable sources like wind and solar energy

Ionic liquid electrolytes explored for rechargeable batteries' advancement; future IL development discussed.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service