Skip to Content
Merck
All Photos(1)

Documents

E49304

Sigma-Aldrich

4-Ethyl-3-thiosemicarbazide

97%

Synonym(s):

1-Amino-3-ethylthiourea, 3-Amino-1-ethylthiourea, 4-Ethylthiosemicarbazide, 4-Ethylthiosemicarbizide, 4-N-Ethyl-3-thiosemicarbazide, 4-N-Ethylthiosemicarbazide, Ethylthiosemicarbazide, N-Ethylhydrazinecarbothioamide, N-Ethylthiosemicarbazide, N1-Ethylthiosemicarbazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5NHCSNHNH2
CAS Number:
Molecular Weight:
119.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

82-84 °C (lit.)

SMILES string

CCNC(=S)NN

InChI

1S/C3H9N3S/c1-2-5-3(7)6-4/h2,4H2,1H3,(H2,5,6,7)

InChI key

SKYYTGUCWARUCL-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tara F Tahir et al.
Sensors (Basel, Switzerland), 12(11), 14968-14982 (2012-12-04)
A flow injection analysis (FIA) incorporating a thiosemicarbazone-based coated wire electrode (CWE) was developed method for the determination of mercury(II). A 0.1 M KNO(3) carrier stream with pH between 1 and 5 and flow rate of 1 mL·min(-1) were used
Carolina G Oliveira et al.
Dalton transactions (Cambridge, England : 2003), 49(28), 9595-9604 (2020-07-01)
Cyclometallated palladium(ii) and platinum(ii) pyrenyl-derived thiosemicarbazone (H2PrR) complexes of the type [M4(μ-S-PrR-κ3-C,N,S)4] (M = PdII, PtII; R = ethyl, cyclohexyl) have been synthesised in good yields and fully characterised. X-ray crystallography showed that the tetranuclear complex [Pt4(μ-S-PrCh-κ3-C,N,S)4](CH3)2COCHCl3 contains an eight-membered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service