Skip to Content
Merck
All Photos(1)

Key Documents

711462

Sigma-Aldrich

o-Xylylenediamine dihydrochloride

≥98%

Synonym(s):

α,α′-Diamino-o-xylene dihydrochloride, 1,2-Bis(aminomethyl)benzene dihydrochloride, ortho-Phenylene-bis(methylamine) dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$458.00

$458.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$458.00

About This Item

Empirical Formula (Hill Notation):
C8H12N2 · 2HCl
CAS Number:
Molecular Weight:
209.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$458.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Assay

≥98%
98.0-102.0% (calculated on dry substance, AT)

form

crystals

impurities

~4% water

functional group

amine

storage temp.

2-8°C

SMILES string

Cl.Cl.NCc1ccccc1CN

InChI

1S/C8H12N2.2ClH/c9-5-7-3-1-2-4-8(7)6-10;;/h1-4H,5-6,9-10H2;2*1H

InChI key

VDEOIFDDSWXYDA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthony P Green et al.
Angewandte Chemie (International ed. in English), 53(40), 10714-10717 (2014-08-21)
The widespread application of ω-transaminases as biocatalysts for chiral amine synthesis has been hampered by fundamental challenges, including unfavorable equilibrium positions and product inhibition. Herein, an efficient process that allows reactions to proceed in high conversion in the absence of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service