Skip to Content
Merck
All Photos(1)

Key Documents

690015

Sigma-Aldrich

o-Xylylenediamine

95%

Synonym(s):

α,α′-Diamino-o-xylene, 1,2-Bis(aminomethyl)benzene, ortho-Phenylene-bis(methylamine)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$655.00

$655.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$655.00

About This Item

Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$655.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

≥94.0% (GC)
95%

form

liquid

functional group

amine

storage temp.

2-8°C

SMILES string

NCc1ccccc1CN

InChI

1S/C8H12N2/c9-5-7-3-1-2-4-8(7)6-10/h1-4H,5-6,9-10H2

InChI key

GKXVJHDEWHKBFH-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service