Skip to Content
Merck
All Photos(2)

Key Documents

279633

Sigma-Aldrich

p-Xylylenediamine

99%

Synonym(s):

1,4-Bis(aminomethyl)benzene, α,α′-Diamino-p-xylene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$157.00
25 G
$561.00

$157.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$157.00
25 G
$561.00

About This Item

Linear Formula:
C6H4(CH2NH2)2
CAS Number:
Molecular Weight:
136.19
Beilstein:
2206190
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$157.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

solid

bp

230 °C/10 mmHg (lit.)

mp

60-63 °C (lit.)

SMILES string

NCc1ccc(CN)cc1

InChI

1S/C8H12N2/c9-5-7-1-2-8(6-10)4-3-7/h1-4H,5-6,9-10H2

InChI key

ISKQADXMHQSTHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-Xylylenediamine (p-XDA) is an aromatic diamine monomer characterized by its two primary amino groups and rigid aromatic structure, which contribute to its exceptional thermal stability and mechanical strength. Its reactivity allows for versatile functionalization and effective cross-linking, making it a suitable building block in polymer synthesis. p-XDA′s solubility in organic solvents facilitates its incorporation into various formulations, including hydrogels for drug delivery systems.

Application

p-Xylylenediamine can be used:
  • As a monomer to synthesize ultra heat-resistant thermoplastic polyamide elastomers suitable for high-temperature applications. It contributes to increased tensile strength, toughness, and Young′s modulus of the elastomer, thereby enhancing its overall mechanical strength.
  • As a cross-linking agent to prepare solvent resistant high performance nanofiltration membranes. p-Xylylenediamine facilitates superior separation performance and stability for N,N-dimethylformamide (DMF) solution.
  • As a dendrimer core to synthesize stimuli responsive nanocarriers for cancer drug delivery.
  • As a capping agent to prepare fluorescent organic–inorganic hybrid CH3NH3PbBr3 nanocrystals to fabricate 3D-printed paper-based microfluidic optical sensor for naked-eye detection of picric acid.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ammara Ejaz et al.
Journal of colloid and interface science, 570, 322-331 (2020-03-15)
Solvent properties such as surface tension, dielectric constant, and viscosity have been extensively studied over more than 150 years to understand their influence on the growth kinetics of nanostructures. Interestingly, these nanoparticles-based studies have missed the influence of solvent molecular geometry.
Weiqiang Zhan et al.
Journal of medicinal chemistry, 50(23), 5655-5664 (2007-10-26)
In light of a proposed molecular mechanism for the C-X-C chemokine receptor type 4 (CXCR4) antagonist 1 (AMD3100), a template with the general structure 2 was designed, and 15 was identified as a lead by means of an affinity binding
Choon Young Lee et al.
Bioorganic & medicinal chemistry letters, 19(22), 6326-6330 (2009-10-14)
Three dendritic polyphenols (generation 1) were synthesized: a syringaldehyde-based dendrimer (1), a vanillin-based dendrimer (2), and an iodinated vanillin-based dendrimer (3). They all showed strong antioxidant activity according to the 1,1-diphenyl-2-picrylhydrazyl (DPPH) radical assay. The syringaldehyde dendrimer was twice and
Francesc Yraola et al.
Journal of medicinal chemistry, 49(21), 6197-6208 (2006-10-13)
Structure activity relationships for semicarbazide-sensitive amine oxidase/vascular adhesion protein-1 (SSAO/VAP-1) were studied using a library of arylalkylamine substrates, with the aim of contributing to the discovery of more efficient SSAO substrates. Experimental data were contrasted with computational docking studies, thereby

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service