Skip to Content
Merck
All Photos(1)

Key Documents

709492

Sigma-Aldrich

(1S,4S,8S)-5-Benzyl-2-isobutyl-8-methoxy-1,8-dimethylbicyclo[2.2.2]octa-2,5-diene

97%

Synonym(s):

(1S,4S,8S) Carreira DOLEFIN Ligand, (1S,4S,8S)--5-Benzyl-8-methoxy-1,8-dimethyl-2-(2′-methylpropyl)-bicyclo[2.2.2]octa-2,5-diene, (1S,4S,8S)-5-Benzyl-2-isobutyl-8-methoxy-1,8-dimethyl-2-bicyclo[2.2.2]octa-2,5-diene

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$431.00

$431.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$431.00

About This Item

Empirical Formula (Hill Notation):
C22H30O
CAS Number:
Molecular Weight:
310.47
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.06

$431.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

optical activity

[α]/D −84±5°, c = 0.5 in chloroform

reaction suitability

reaction type: click chemistry

refractive index

n20/D 1.522

functional group

ether
phenyl

SMILES string

CO[C@@]1(C)C[C@@]2(C)C=C(Cc3ccccc3)[C@@H]1C=C2CC(C)C

InChI

1S/C22H30O/c1-16(2)11-19-13-20-18(12-17-9-7-6-8-10-17)14-21(19,3)15-22(20,4)23-5/h6-10,13-14,16,20H,11-12,15H2,1-5H3/t20-,21+,22-/m0/s1

InChI key

ACWLDJOHMGJACE-BDTNDASRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(1S,4S,8S)-5-Benzyl-2-isobutyl-8-methoxy-1,8-dimethylbicyclo[2.2.2]octa-2,5-diene, an enantiopure diene ligand developed by Carreira, is most commonly used in asymmetric catalysis. It is generally prepared from commercially available (R)-carvone.[1]

Application

(1S,4S,8S)-5-Benzyl-2-isobutyl-8-methoxy-1,8-dimethylbicyclo[2.2.2]octa-2,5-diene is mainly useful for the stereoselective synthesis of diarylmethine stereogenic centers.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhodium-catalyzed enantioselective conjugate addition of organoboronic acids to alpha, beta-unsaturated sulfones
Mauleon P and Carretero JC
Organic Letters, 6(18), 3195-3198 (2004)
(1S,4S,8S)?8?Methoxy?1,8?dimethyl?2?(2?methylpropyl)?5?(phenylmethyl)bicyclo[2.2.2]octa?2,5?diene
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2011)

Related Content

Carreira Research & SpiroChem innovate spirocyclic building blocks, enhancing drug discovery with diverse functionalization sites and compact modules.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service