Skip to Content
Merck
All Photos(1)

Key Documents

656763

Sigma-Aldrich

Di-μ-chlorotetraethylene dirhodium(I)

Synonym(s):

Bis[chlorobis(ethylene)rhodium], Bis[chlorodiethylenerhodium], Chlorobis(ethylene)rhodium dimer, Dichlorotetra(ethene)dirhodium, Dichlorotetrakis(ethene)dirhodium, Diethylenerhodium chloride dimer, Divinylrhodium chloride dimer, Tetrakis(ethylene)dichlorodirhodium, [Rh(C2H4)2Cl]2

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$162.00
1 G
$891.00

$162.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$162.00
1 G
$891.00

About This Item

Empirical Formula (Hill Notation):
C8H16Cl2Rh2
CAS Number:
Molecular Weight:
388.93
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$162.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

reaction suitability

core: rhodium
reagent type: catalyst

storage temp.

−20°C

SMILES string

Cl[Rh].Cl[Rh].C=C.C=C.C=C.C=C

InChI

1S/4C2H4.2ClH.2Rh/c4*1-2;;;;/h4*1-2H2;2*1H;;/q;;;;;;2*+1/p-2

InChI key

OOQJCPOXJFCGCR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Di-μ-chlorotetraethylene dirhodium(I) may be used in the preparation of Π-bound rhodium organometallic complexes of ROMP-derived poly(1,5-cyclooctadiene) (poly(COD)). [ROMP= Ring-Opening Metathesis Polymerization; COD= 1,5-cyclooctadiene][1]
Precursor to Rh(I) norbornadiene catalyst used in an asymmetric 1,4-addition of arylboronic acids to cyclic enones.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polycyclooctadiene Complexes of Rhodium (I): Direct Access to Organometallic Nanoparticles.
Mavila S, et al.
Angewandte Chemie (International Edition in English), 52(22), 5767-5770 (2013)
Tetrahedron, 63, 12961-12961 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service