Skip to Content
Merck
All Photos(1)

Documents

708704

Sigma-Aldrich

RuCl[(R,R)-FsDPEN](p-cymene)

90%

Synonym(s):

[N-[(1R,2R)-2-(Amino-κN)-1,2-diphenylethyl]-2,3,4,5,6-pentafluorobenzenesulfonamidato-κN]chloro[(1,2,3,4,5,6-η)-1-methyl-4-(1-methylethyl)benzene]-ruthenium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H28ClF5N2O2RuS
CAS Number:
Molecular Weight:
712.14
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

90%

form

powder

optical activity

[α]25/D −16°, c = 0.1 in chloroform

mp

238-244 °C

storage temp.

2-8°C

SMILES string

CC(C)c1ccc(C)cc1.N[C@@H]([C@H](N([Ru]Cl)S(=O)(=O)c2c(F)c(F)c(F)c(F)c2F)c3ccccc3)c4ccccc4

InChI

1S/C20H14F5N2O2S.C10H14.ClH.Ru/c21-13-14(22)16(24)20(17(25)15(13)23)30(28,29)27-19(12-9-5-2-6-10-12)18(26)11-7-3-1-4-8-11;1-8(2)10-6-4-9(3)5-7-10;;/h1-10,18-19H,26H2;4-8H,1-3H3;1H;/q-1;;;+2/p-1/t18-,19-;;;/m1.../s1

InChI key

UWFMZLATRGEOIW-ZJPTYAPPSA-M

Legal Information

Sold in collaboration with Takasago for research purposes only. WO9720789

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>212.0 °F - open cup

Flash Point(C)

> 100 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service