Skip to Content
Merck
All Photos(1)

Key Documents

703907

Sigma-Aldrich

RuCl(p-cymene)[(R,R)-Ts-DPEN]

Synonym(s):

[N-[(1R,2R)-2-(Amino-κN)-1,2-diphenylethyl]-4-methylbenzenesulfonamidato-κN]chloro[(1,2,3,4,5,6-η)-1-methyl-4-(1-methylethyl)benzene]-ruthenium

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$36.50
500 MG
$132.00

$36.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$36.50
500 MG
$132.00

About This Item

Empirical Formula (Hill Notation):
C31H35ClN2O2RuS
CAS Number:
Molecular Weight:
636.21
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$36.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

optical activity

[α]20/D −116°, c = 0.1 in chloroform

mp

215 °C

functional group

amine
phenyl

storage temp.

2-8°C

SMILES string

CC(C)c1ccc(C)cc1.Cc2ccc(cc2)S(=O)(=O)N([Ru]Cl)[C@@H]([C@H](N)c3ccccc3)c4ccccc4

InChI

1S/C21H21N2O2S.C10H14.ClH.Ru/c1-16-12-14-19(15-13-16)26(24,25)23-21(18-10-6-3-7-11-18)20(22)17-8-4-2-5-9-17;1-8(2)10-6-4-9(3)5-7-10;;/h2-15,20-21H,22H2,1H3;4-8H,1-3H3;1H;/q-1;;;+2/p-1/t20-,21-;;;/m1.../s1

InChI key

AZFNGPAYDKGCRB-AGEKDOICSA-M

Looking for similar products? Visit Product Comparison Guide

General description

RuCl(p-cymene)[(R,R)-Ts-DPEN] is a chiral diamine ligand complexed with ruthenium.[1]

Application

Phosphine cationic ruthenium catalysts used for enantioselective hydrogenation of:
  • Quinolines
  • N-Alkyl ketimines
  • Antitumor and antiproliferative derivatives of natural products isolated from bacteria
  • Hydroxy arylaldehydes via Rap-Stoermer-enantioselective transfer hydrogenation

Catalyst with improved performance modified via microenvironment engineering of nanocages

Legal Information

Sold in collaboration with Takasago for research purposes only. WO9720789

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbonyl compounds: still central to organic synthesis
Aldrichimica Acta, 41(4) (2008)

Articles

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service