Skip to Content
Merck
All Photos(2)

Key Documents

707961

Sigma-Aldrich

Grubbs Catalyst® C833

95%

Synonym(s):

Dichloro(PCy3)[(tricyclohexylphosphoranyl)methylidene]Ru tetrafluoroborate, Dichloro(tricyclohexylphosphine)[(tricyclohexylphosphoranyl)methylidene]ruthenium(II) tetrafluoroborate, Piers 1st generation metathesis catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H67BCl2F4P2Ru
CAS Number:
Molecular Weight:
832.66
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst
reaction type: Ring Opening Metathesis Polymerisation

mp

203 °C (dec.)

storage temp.

2-8°C

SMILES string

F[B-](F)(F)F.C1CCC(CC1)P(C2CCCCC2)C3CCCCC3.Cl\[Ru](Cl)=C/[P+](C4CCCCC4)(C5CCCCC5)C6CCCCC6

InChI

1S/C19H34P.C18H33P.BF4.2ClH.Ru/c1-20(17-11-5-2-6-12-17,18-13-7-3-8-14-18)19-15-9-4-10-16-19;1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;2-1(3,4)5;;;/h1,17-19H,2-16H2;16-18H,1-15H2;;2*1H;/q+1;;-1;;;+2/p-2

InChI key

YFCZUTDHPBTFQL-UHFFFAOYSA-L

Application

Cationic 4-coordinate catalyst is considered “pre-activated” because no ligand dissociation is required for formation of the active catalytic species. Has reactivity profile similar to Grubbs Catalyst® 1st Generation but is active at much lower temperatures (below 0 °C).

Legal Information

Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Romero, P. E.; Piers, W. E.; McDonald, R.
Angewandte Chemie (International Edition in English), 6161-6161 (2004)
Kinetic and thermodynamic analysis of processes relevant to initiation of olefin metathesis by ruthenium phosphonium alkylidene catalysts.
Leitao EM, et al.
Journal of the American Chemical Society, 132(8), 2784-2794 (2010)

Articles

Alkyne metathesis has been a useful tool for C–C bond formation since the discovery of structurally well-defined metal alkylidynes by Schrock and coworkers.

Related Content

Research focuses on ruthenium alkylidene complexes for versatile olefin metatheses catalysis in the Grubbs group.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service