Skip to Content
Merck
All Photos(1)

Documents

707716

Sigma-Aldrich

N-Methyl-13C,d3-3-piperidinol

99 atom % 13C, 98 atom % D, 98% (CP)

Synonym(s):

1-Methyl-13C,d3-3-piperidinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C513CH10D3NO
CAS Number:
Molecular Weight:
119.18
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % D

Assay

98% (CP)

form

liquid

bp

76-78 °C/11 mmHg

density

1.033 g/mL at 25 °C

mass shift

M+4

SMILES string

[2H][13C]([2H])([2H])N1CCCC(O)C1

InChI

1S/C6H13NO/c1-7-4-2-3-6(8)5-7/h6,8H,2-5H2,1H3/i1+1D3

InChI key

UKANCZCEGQDKGF-KQORAOOSSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F

Flash Point(C)

70 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service