Skip to Content
Merck
All Photos(1)

Documents

107778

Sigma-Aldrich

Allyltrichlorosilane

95%

Synonym(s):

Propen-3-yltrichlorosilane, Trichloro(2-propenyl)silane, Trichloro-2-propen-1-ylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2SiCl3
CAS Number:
Molecular Weight:
175.52
Beilstein:
1743449
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

vapor pressure

10 mmHg ( 16.1 °C)

Assay

95%

refractive index

n20/D 1.443 (lit.)

bp

116 °C/750 mmHg (lit.)

density

1.211 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Cl[Si](Cl)(Cl)CC=C

InChI

1S/C3H5Cl3Si/c1-2-3-7(4,5)6/h2H,1,3H2

InChI key

HKFSBKQQYCMCKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Allyltrichlorosilane is used as a key building block for the synthesis of various organosilicon compounds. It has a wide range of applications across various industries, including the production of silicone rubber, resins, coatings, and adhesives. In organic synthesis, allyltrichlorosilane undergoes various chemical reactions, including hydrolysis, cross-coupling reactions, hydrosilylation, allylation of aldehydes and ketones, and palladium-catalyzed allylic substitutions.

Application

Allyltrichlorosilane can be used as a reactant to synthesize poly(hydrosilane)s via free radical catalyzed polymerization reaction.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Núria Torras et al.
Soft matter, 13(40), 7264-7272 (2017-09-28)
A new temperature-controlled smart soft material micropillar array has been fabricated via in situ integration of the liquid-crystalline elastomer-based component into the hybrid microdevice. Such design allows for developing pushing elements with fast lifetime values of ca. 5 s, and
Robert Foreman et al.
Molecular systems biology, 16(2), e9146-e9146 (2020-02-12)
Gene expression variability in mammalian systems plays an important role in physiological and pathophysiological conditions. This variability can come from differential regulation related to cell state (extrinsic) and allele-specific transcriptional bursting (intrinsic). Yet, the relative contribution of these two distinct

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service