Skip to Content
Merck
All Photos(1)

Key Documents

702897

SAFC

1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene

97%

Synonym(s):

1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene, SIMes

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$34.00
500 MG
$172.00

$34.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest a Sample

Select a Size

Change View
100 MG
$34.00
500 MG
$172.00

About This Item

Empirical Formula (Hill Notation):
C21H26N2
CAS Number:
Molecular Weight:
306.44
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$34.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest a Sample

Assay

97%

form

solid

mp

85-100 °C

storage temp.

2-8°C

SMILES string

Cc1cc(C)c(N2[C]N(CC2)c3c(C)cc(C)cc3C)c(C)c1

InChI

1S/C21H26N2/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6/h9-12H,7-8H2,1-6H3

InChI key

LSMWOQFDLBIYPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Mei et al.
BMC genomics, 16, 336-336 (2015-04-23)
Genetic heritability and expression study have shown that different diabetes traits have common genetic components and pathways. A computationally efficient pathway analysis of GWAS results will benefit post-GWAS study of SNP associations and identification of common genetic pathways from diabetes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service