Skip to Content
Merck
All Photos(2)

Key Documents

230227

Sigma-Aldrich

Tris(4-bromophenyl)ammoniumyl hexachloroantimonate

technical grade

Synonym(s):

Hexachloroantimonate tris(4-bromophenyl)amine, Magic Blue, Tris(4-bromophenyl)aminium hexachloroantimonate, Tris(p-bromophenyl)aminium hexachloroantimonate(1-), Tris(p-bromophenyl)aminium hexachloroantimonate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$28.60
5 G
$132.00

$28.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$28.60
5 G
$132.00

About This Item

Linear Formula:
(BrC6H4)3NSbCl6
CAS Number:
Molecular Weight:
816.48
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$28.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

form

powder

mp

142 °C (dec.) (lit.)

SMILES string

Cl[Sb-](Cl)(Cl)(Cl)(Cl)Cl.Brc1ccc(cc1)[N+](c2ccc(Br)cc2)c3ccc(Br)cc3

InChI

1S/C18H12Br3N.6ClH.Sb/c19-13-1-7-16(8-2-13)22(17-9-3-14(20)4-10-17)18-11-5-15(21)6-12-18;;;;;;;/h1-12H;6*1H;/q+1;;;;;;;+5/p-6

InChI key

SDHBPVANTRLAKE-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Application

Tris(4-bromophenyl)ammoniumyl hexachloroantimonate can be used as a strong oxidant for the chemical doping of conjugated polymers[1]. It can also be used as a catalyst for the deprotection of tert-butyldimethylsilyl and tetrahydropyranyl ethers[2].

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chemical doping of conjugated polymers with the strong oxidant magic blue
Hofmann A, et al.
Advanced Electronic Materials, 6, 2000249-2000249 (2020)
A mild and efficient cleavage of tert-butyldimethylsilyl (TBS) and tetrahydropyranyl (THP) ethers using a catalytic amount of TBPA+ SbCl6
Xu Y, et al.
Tetrahedron Letters, 49, 3634-3637 (2008)
Rachel L Meyer et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(44), 9905-9914 (2020-03-21)
The rational control of the electrochemical properties of polyoxovanadate-alkoxide clusters is dependent on understanding the influence of various synthetic modifications on the overall redox processes of these systems. In this work, the electronic consequences of ligand substitution at the heteroion
Hong Zhang et al.
Analytica chimica acta, 1107, 107-112 (2020-03-24)
Many important chemical transformations proceed by way of ionic and/or neutral intermediates. Great effort has been expended to understand the mechanism, with only minimum attention given to separate associated ionic and neutral intermediates. Herein, we present a nebulization method followed

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service