702706
Sodium tert-butoxide solution
2 M in THF, ZerO2®
Synonym(s):
Degassed and low oxygen sodium tert-butoxide solution
About This Item
Recommended Products
grade
ZerO2®
Quality Level
form
liquid
concentration
2 M in THF
refractive index
n20/D 1.413
density
0.911 g/mL at 25 °C
SMILES string
[Na+].CC(C)(C)[O-]
InChI
1S/C4H9O.Na/c1-4(2,3)5;/h1-3H3;/q-1;+1
InChI key
MFRIHAYPQRLWNB-UHFFFAOYSA-N
Related Categories
Application
Packaging
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Self-heat. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
WGK 1
Flash Point(F)
-4.0 °F - closed cup
Flash Point(C)
-20 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service