Skip to Content
Merck
All Photos(1)

Key Documents

8.04918

Sigma-Aldrich

Potassium tert-butylate

for synthesis

Synonym(s):

Potassium tert-butylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COK
CAS Number:
Molecular Weight:
112.21
MDL number:
UNSPSC Code:
12352107
EC Index Number:
212-740-3
NACRES:
NA.22
grade:
for synthesis
synthesis grade
form:
powder
Pricing and availability is not currently available.

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

0.02 hPa ( 20 °C)

Assay

≥98.0% (acidimetric)

form

powder

autoignition temp.

483 °C

pH

13 (20 °C, 5 g/L in H2O, Hydrolysis)

mp

256-258 °C

density

1.19 g/cm3 at 20 °C

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

[K+].[O-]C(C)(C)C

InChI

1S/C4H9O.K/c1-4(2,3)5;/h1-3H3;/q-1;+1

InChI key

LPNYRYFBWFDTMA-UHFFFAOYSA-N

General description

Potassium tert-butylate (Potassium tert-Butoxide) is a strong, non-nucleophilic alkoxide base in organic chemistry. Potassium tert-butoxide catalyzes the reaction of hydro silanes and heterocyclic compounds to generate the silyl derivatives. It is used to catalyze the Michael addition reaction, Ramberg-Baecklund reaction, Pinacol rearrangement.[1]

Application

Potassium tert-butylate is used to catalyzes the Michael addition reaction.[2]

Analysis Note

Assay (acidimetric): ≥ 98,0 %(m)
Free KOH: ≤ 1 %(m)
Identity (IR): conforms

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Self-heat. 2 - Skin Corr. 1A - Water-react 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service