Skip to Content
Merck
All Photos(3)

Key Documents

702757

Sigma-Aldrich

Potassium tert-pentoxide solution

2 M in THF

Synonym(s):

Potassium tert-amylate, Potassium (2-methyl-2-butoxide), Potassium tert-pentylate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$387.00
500 ML
$1,890.00

$387.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$387.00
500 ML
$1,890.00

About This Item

Linear Formula:
CH3CH2C(CH3)2OK
CAS Number:
Molecular Weight:
126.24
Beilstein:
4149025
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

$387.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

form

liquid

concentration

2 M in THF

density

0.920 g/mL at 25 °C

SMILES string

[K+].CCC(C)(C)[O-]

InChI

1S/C5H11O.K/c1-4-5(2,3)6;/h4H2,1-3H3;/q-1;+1

InChI key

ZRLVQFQTCMUIRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassiumtert-pentoxide solution is a strong and sterically hindered alkoxide baseprepared by treating oil-free potassium hydride with anhydrous t-pentylalcohol in anhydrous solvent.

Application

Potassium tert-pentoxide solution can be used as a base in the:
  • Wittig reaction of ketoesters with methyltriphenylphosphonium bromide to synthesize alkenes.
  • Palladium-catalyzed Buchwald-Hartwig amination reaction.
It can also be used:
  • To prepare potassium tert-pentoxide-crown ether complexes, which is used as a Lewis base catalyst in Mukaiyama aldol reaction.
  • As an activator for (difluoromethyl)trimethylsilane to generate difluoromethyl-carbanion which reacts with isothiocyanate to synthesize α, α-difluoromethyl thioamide.

Features and Benefits

In the absence of air and moisture, potassium tert-pentoxide solutions in benzene or toluene may be stored in a refrigerator for up to 1 year without significant deterioration.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - Solvent

Flash Point(C)

-17 °C - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service