Skip to Content
Merck
All Photos(1)

Key Documents

678201

Sigma-Aldrich

Allyl methyl ether

97%

Synonym(s):

3-Methoxy-1-propene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2OCH3
CAS Number:
Molecular Weight:
72.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.3803

bp

42-43 °C/760 mmHg

density

0.768 g/mL at 25 °C

functional group

ether

storage temp.

2-8°C

SMILES string

COCC=C

InChI

1S/C4H8O/c1-3-4-5-2/h3H,1,4H2,2H3

InChI key

FASUFOTUSHAIHG-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

<-20.0 °F - closed cup

Flash Point(C)

< -28.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service