Skip to Content
Merck
All Photos(5)

Key Documents

C6645

Sigma-Aldrich

Cytosine β-D-arabinofuranoside hydrochloride

crystalline

Synonym(s):

1-(β-D-Arabino­furanosyl)­cytosine hydrochloride, Ara-C hydrochloride, Arabinocytidine hydrochloride, Arabinosylcytosine hydrochloride, Cytarabine hydrochloride, Cytosine arabinoside hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N3O5 · HCl
CAS Number:
Molecular Weight:
279.68
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (inorganic)

Assay

≥99% (HPLC)

form

crystalline

mp

197-198 °C (lit.)

storage temp.

2-8°C

SMILES string

Cl[H].NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)[C@@H]2O

InChI

1S/C9H13N3O5.ClH/c10-5-1-2-12(9(16)11-5)8-7(15)6(14)4(3-13)17-8;/h1-2,4,6-8,13-15H,3H2,(H2,10,11,16);1H/t4-,6-,7+,8-;/m1./s1

InChI key

KCURWTAZOZXKSJ-JBMRGDGGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cytosine β-D-arabinofuranoside hydrochloride has been used in fetal bovine serum supplemented Dulbecco′s modified eagle medium (FBS-DMEM) and B27/neurobasal-A medium to inhibit the growth of glial cells. It has also been used to inhibit astrocyte proliferation in embryonic spinal cord neurons.

Biochem/physiol Actions

Ara-C is phosphorylated to Ara-CTP and is incorporatee into DNA. It inhibits DNA replication by forming cleavage complexes with topoisomerase I resulting in DNA fragmentation, and ultimately induces apoptosis via the PKC signaling pathway. Does not inhibit RNA synthesis. Anti-leukemia agent.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Muta. 2 - Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

JNK plays a key role in tau hyperphosphorylation in Alzheimer's disease models
Ploia C, et al.
Journal of Alzheimer'S Disease, 26(2), 315-329 (2011)
Vindhya Vijay et al.
Leukemia research, 84, 106180-106180 (2019-07-13)
One of the greatest challenges in treating acute myeloid leukemia (AML) is chemotherapy refractory disease. Previously, we demonstrated a novel mechanism whereby AML-induced endothelial cell (EC) activation leads to subsequent leukemia cell adherence, quiescence and chemoresistance, identifying activated ECs as
Reduced activity of AMP-activated protein kinase protects against genetic models of motor neuron disease
Lim MA, et al.
The Journal of Neuroscience, 32(3), 1123-1141 (2012)
Joost J Leenders et al.
The Journal of biological chemistry, 285(35), 27449-27456 (2010-06-23)
Pathological forms of left ventricular hypertrophy (LVH) often progress to heart failure. Specific transcription factors have been identified that activate the gene program to induce pathological forms of LVH. It is likely that apart from activating transcriptional inducers of LVH
Julie M Vose et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(13), 1662-1668 (2013-03-13)
This clinical trial evaluated standard-dose radioimmunotherapy with a chemotherapy-based transplantation regimen followed by autologous hematopoietic cell transplantation versus rituximab with the same regimen in patients with relapsed diffuse large B-cell lymphoma (DLBCL). Patients with chemotherapy-sensitive persistent or relapsed DLBCL were

Articles

DNA damage and repair mechanism is vital for maintaining DNA integrity. Damage to cellular DNA is involved in mutagenesis, the development of cancer among others.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service