Skip to Content
Merck
All Photos(1)

Key Documents

112046

Sigma-Aldrich

2-Methyl-2-propen-1-ol

98%

Synonym(s):

β-Methallyl alcohol, ‘Isobutenol’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)CH2OH
CAS Number:
Molecular Weight:
72.11
Beilstein:
969226
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

refractive index

n20/D 1.426 (lit.)

bp

113-115 °C (lit.)

density

0.857 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(=C)CO

InChI

1S/C4H8O/c1-4(2)3-5/h5H,1,3H2,2H3

InChI key

BYDRTKVGBRTTIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The rate coefficient for gas-phase reaction of 2-Methyl-2-propen-1-ol (MePro) with OH radical can be determined by the relative rate method or by using the pulsed laser photolysis-laser induced fluorescence technique[1].

Application

2-Methyl-2-propen-1-ol (MePro) attaches to hemicellulose acetylated galactoglucomannan (AcGGM) backbone providing it with pendant sites that allow subsequent cross-linking and hydrogel formation[2].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Alkenyl-Functionalized Precursors for Renewable Hydrogels Design.
Journal of Polymer Science Part A: Polymer Chemistry, 47(17), 3595-3606 (2009)
Pablo M Cometto et al.
The journal of physical chemistry. A, 112(19), 4444-4450 (2008-04-25)
Rate coefficients for the gas-phase reactions of OH radicals with four unsaturated alcohols, 3-methyl-3-buten-1-ol (k1), 2-buten-1-ol (k2), 2-methyl-2-propen-1-ol (k3) and 3-buten-1-ol (k4), were measured using two different techniques, a conventional relative rate method and the pulsed laser photolysis-laser induced fluorescence
R Bittman et al.
Chemistry and physics of lipids, 50(2), 99-103 (1989-05-01)
The synthesis of C2-methyl-1,2-di-O-hexadecylglycerophosphocholine from methylallyl alcohol is described and the 13C-NMR assignments of all glycerol backbone and headgroup resonances are provided. Bond rotations in the glycerol backbone of this diether phospholipid derivative are impeded by steric hindrance exerted by

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service