Skip to Content
Merck
All Photos(1)

Key Documents

670936

Sigma-Aldrich

O-(2-Carboxyethyl)polyethylene glycol

5,000

Synonym(s):

Polyethylene glycol, Polyethylene glycol 3400 monoacid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2CH2O)nCH2CH2COOH
CAS Number:
MDL number:
UNSPSC Code:
51171641
NACRES:
NA.22
Pricing and availability is not currently available.

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service