Skip to Content
Merck
All Photos(1)

Key Documents

920401

Sigma-Aldrich

Methoxypolyethylene glycol

average MN 1,000 (by NMR), (Propionic acid), methoxy

Synonym(s):

O-(2-Carboxyethyl)-O′-methylpolyethylene glycol 1,000, PEG acid, PEG carboxylic acid, Polyethylene glycol, mPEG-COOH, mono-Methyl polyethylene glycol 1′000 propionic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$74.10

$74.10


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$74.10

About This Item

Linear Formula:
CH3(OCH2CH2)nOCH2CH2COOH
UNSPSC Code:
12352106
NACRES:
NA.23

$74.10


Estimated to ship onMay 25, 2025


Request a Bulk Order

Product Name

Methoxypolyethylene glycol 1,000 propionic acid,

form

solid

Quality Level

mol wt

average Mn 1,000 (by NMR)

color

white to off-white

Ω-end

(Propionic acid)

α-end

methoxy

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

2-8°C

Application

Methoxypolyethylene glycol 1,000 propionic acid is a pegylation agent used to conjugate proteins to reduce immunogenicity and prolong serum half-life of proteins and enzymes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Fajdetić et al.
European journal of medicinal chemistry, 46(8), 3388-3397 (2011-05-24)
Synthesis, antibacterial activity and pharmacokinetic properties of a novel class of macrolide antibiotics-macrolones-derived from azithromycin, comprising oxygen atom(s) in the linker and either free or esterified quinolone 3-carboxylic group, are reported. Selected compounds showed excellent antibacterial potency towards key erythromycin
A Ton J Dirks et al.
Bioconjugate chemistry, 20(6), 1129-1138 (2009-05-21)
The Cu(I)-catalyzed azide-alkyne cycloaddition (CuAAC) has recently proven to be a powerful synthetic tool in various fields of chemistry, including protein-polymer conjugation. In this article, we describe a fluorogenic CuAAC, which allows for efficient monitoring of protein-polymer conjugation. We show

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service