Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
O-(2-Carboxyethyl)polyethylene glycol
Synonym(s):
Polyethylene glycol 3400 monoacid
Linear Formula:
HO(CH
2
CH
2
O)
n
CH
2
CH
2
COOH
CAS Number:
117786-94-4
Products
Brand
Sigma-Aldrich (2)
Functional Group
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
670812
O-(2-Carboxyethyl)polyethylene glycol,
3,000
Expand
View Pricing
670936
O-(2-Carboxyethyl)polyethylene glycol,
5,000
Expand
View Pricing