Skip to Content
Merck
All Photos(3)

Key Documents

634204

Sigma-Aldrich

2-Amino-5-bromo-4,6-dimethylpyridine

97%

Synonym(s):

5-Bromo-4,6-dimethyl-2-pyridinamine, 6-Amino-3-bromo-2,4-lutidine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$208.00

$208.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$208.00

About This Item

Empirical Formula (Hill Notation):
C7H9BrN2
CAS Number:
Molecular Weight:
201.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$208.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Assay

97%

form

solid

mp

144-148 °C (lit.)

functional group

bromo

SMILES string

Cc1cc(N)nc(C)c1Br

InChI

1S/C7H9BrN2/c1-4-3-6(9)10-5(2)7(4)8/h3H,1-2H3,(H2,9,10)

InChI key

BFRMYMFNSHASRJ-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service